

2,2,3,4,4,4-Hexafluorobutyl methacrylate 96% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36405-47-7
:
Formula:C₈H₈F₆O₂
MDL Number:MFCD00042311
Boiling Pt:158 °C
Density:1,352
Flash Pt:56 °C (132 °F)
UN:3272
ADR:3,III
:250.14 g/mol
:GHS02
:GHS07
Related Information
Frequently Bought Together