(3-Carboxypropyl)triphenylphosphonium bromide 97%
: 3-Carboxypropyltriphenylphosphonium bromide
:17857-14-6
:
:C₂₂H₂₂BrO₂P
:MFCD00274196
:429.30 g/mol
:247…249 °C
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:White Powder and/or granules
- Infrared spectrum:Conforms
- Melting point:243 to 250 °C
- Titration Argentometric:≥96.0 %
Frequently Bought Together