

Dysprosium(III) perchlorate hexahydrate 50% (w/w) in aqueous solution, Reagent Grade
Catalog # 40574.NA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:2 g
- Seq:3107
Specifications
About this item
CAS Number:14692-17-2
:
Formula:Cl₃DyO₁₂·6H₂O
MDL Number:MFCD00150227
:568.94 g/mol
Storage Temperature:Ambient
EINECS:237-838-3
UN:3211
ADR:5.1,II
:GHS03
:GHS07