
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:175136-62-6
:
Formula:C₂₄H₉F₁₈P
MDL Number:MFCD00042428
:670.28 g/mol
Boiling Pt:128…130 °C (0,5 mmHg)
Melting Pt:98…102 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together