Potassium acetate 8 M in aqueous solution RNase-free
Catalog # J62856.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 ml
- Seq:2590
Specifications
About this item
:
:98.14 g/mol
Storage Temperature:Ambient
EINECS:204-822-2
CAS Number:127-08-2
Formula:CH₃COOK
MDL Number:MFCD00012458