


4-Chloro-6-nitro-o-toluidine 97%
ACRO326400050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Orange Crystals or needles
- Infrared spectrum:Conforms
- Melting point:126 to 132 °C
- GC:≥96.0 %
:
:186.60 g/mol
:126…132 °C
:Ambient
:6.1,III
:GHS07
:GHS08
:GHS09
:62790-50-5
:C₇H₇ClN₂O₂
:MFCD01320687