2,2,3,3,4,4,4-Heptafluorobutylamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:374-99-2
:
Formula:C₄H₄F₇N
MDL Number:MFCD00014817
:199.07 g/mol
Storage Temperature:Ambient
EINECS:206-780-0
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together