D-(+)-Cellobiose ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:528-50-7
:
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00136034
Merck Index:14,1961
Melting Pt:234 °C (dec.)
:342.30 g/mol
Storage Temperature:Ambient
EINECS:208-436-5
Related Information
Frequently Bought Together