Order Entry
Northern Ireland
ContactUsLinkComponent
(2-Butyl)triphenylphosphonium bromide 96%
Irritant
(2-Butyl)triphenylphosphonium bromide 96%
Catalog # L01532.06
CAS Number:  
Irritant
(2-Butyl)triphenylphosphonium bromide 96%
Catalog # L01532.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :399.32 g/mol
Melting Pt:227…237 °C
EINECS:223-587-7
 :GHS07
Storage Temperature:Ambient
CAS Number:3968-92-1
Formula:C₂₂H₂₄BrP
MDL Number:MFCD00050236