Dysprosium(III) acetate tetrahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15280-55-4
:
Formula:C₆H₉DyO₆·4H₂O
MDL Number:MFCD00013033
:411.68 g/mol
Melting Pt:120 °C (dec.)
Storage Temperature:Ambient
EINECS:242-565-8
Related Information
Frequently Bought Together