5-Chloro-2-(methylamino)benzophenone
Catalog # PROOICRS0199
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:245.71 g/mol
EINECS:213-822-1
CAS Number:1022-13-5
Formula:C₁₄H₁₂ClNO
MDL Number:MFCD00008284