Order Entry
Northern Ireland
ContactUsLinkComponent
5-Pyrimidineboronic acid pinacol ester 95%
Irritant
5-Pyrimidineboronic acid pinacol ester 95%
Catalog # H27147.03
CAS Number:  
Irritant
5-Pyrimidineboronic acid pinacol ester 95%
Catalog # H27147.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :206.05 g/mol
Melting Pt:60…66 °C
 :GHS07
CAS Number:321724-19-0
Formula:C₁₀H₁₅BN₂O₂
MDL Number:MFCD05155223