

Diphenylphosphoryl azide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:26386-88-9
:
Formula:C₁₂H₁₀N₃O₃P
MDL Number:MFCD00001987
:275.20 g/mol
Boiling Pt:161…162 °C (0,5 mmHg)
Density:1,275
Flash Pt:>110 °C (230 °F)
EINECS:247-644-0
UN:3278
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together