ContactUsLinkComponent
3,4-(Methylenedioxy)phenylboronic acid 98%
Irritant
3,4-(Methylenedioxy)phenylboronic acid 98%
Certificates
undefined
Irritant
3,4-(Methylenedioxy)phenylboronic acid 98%
Specification Test Results
  • Appearance:
    White to yellow to light beige Powder
  • Infrared spectrum:
    Conforms
  • Titration with NaOH:
    97.5 to 106.0 %