Order Entry
Northern Ireland
ContactUsLinkComponent
2-(Trimethylsilyl)ethoxymethyl chloride tech. 90% stabilised
Corrosive
Flammable
2-(Trimethylsilyl)ethoxymethyl chloride tech. 90% stabilised
Catalog # H55074.06
CAS Number:  
Corrosive
Flammable
2-(Trimethylsilyl)ethoxymethyl chloride tech. 90% stabilised
Catalog # H55074.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Assay (GC):
    >88.0%

Specification Test Results

About this item

 :
 :166.72 g/mol
Boiling Pt:170…172 °C
Density:0,942
Flash Pt:46 °C (115 °F)
Storage Temperature:Refrigerator
EINECS:278-483-4
UN:2920
ADR:8,II
 :Stabilised with N-ethyldiisopropylamine (N,N-diisopropylethylamine) 0,1%
 :GHS05
 :GHS02
CAS Number:76513-69-4
Formula:C₆H₁₅ClOSi
MDL Number:MFCD00009919