

1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:102488-47-1
:
Formula:C₁₀H₁₀ClF₁₃Si
MDL Number:MFCD00042359
:440.71 g/mol
Boiling Pt:189…191 °C
Density:1,461
Flash Pt:52 °C (125 °F)
Storage Temperature:Ambient
ADR:8,II
:GHS05
:GHS02
Related Information
Frequently Bought Together