Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Theophyllin

Theophyllin

Glass bottle for solids, 500 mg
Catalog # PROOMM0176.00
Theophyllin
Catalog # PROOMM0176.00
CAS Number:  58-55-9
Theophyllin

Glass bottle for solids, 500 mg

CAS Number:  58-55-9
Price
£1,500.00
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline
  • CAS Number:58-55-9
  • Formula:C₇H₈N₄O₂
  • MDL Number:MFCD00079619
  • Molecular Weight:180.16 g/mol
  • EINECS:200-385-7
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.