Theophyllin
Catalog # PROOBP327
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:180.16 g/mol
EINECS:200-385-7
UN:2811
ADR:6.1,III
CAS Number:58-55-9
Formula:C₇H₈N₄O₂
MDL Number:MFCD00079619