Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
Bis(pinacolato)diboron 98%
 
undefined
 
Bis(pinacolato)diboron 98%

 

  • Appearance:
    White Powder or platelets
  • Infrared spectrum:
    Conforms
  • Melting point:
    135 to 140 °C
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together