Order Entry
Northern Ireland
ContactUsLinkComponent
Ergocalciferol (vitamin D2, calciferol)
Ergocalciferol (vitamin D2, calciferol)
Catalog # MOLEM77131888
Supplier:  Molekula
CAS Number:  
Ergocalciferol (vitamin D2, calciferol)
Catalog # MOLEM77131888
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
ADR:6.1,II
 :396.66 g/mol
EINECS:200-014-9
UN:2811
CAS Number:50-14-6
Formula:C₂₈H₄₄O
MDL Number:MFCD00166988