

Ergocalciferol (vitamin D2, calciferol)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:50-14-6
:
Formula:C₂₈H₄₄O
MDL Number:MFCD00166988
Merck Index:14,10018
Melting Pt:114…118 °C
EINECS:200-014-9
UN:2811
ADR:6.1,II
:396.66 g/mol
:GHS06
:GHS08
Related Information
Frequently Bought Together