Order Entry
Northern Ireland
ContactUsLinkComponent
Ergocalciferol (vitamin D2, calciferol) 97+%
Toxic
Health
Ergocalciferol (vitamin D2, calciferol) 97+%
Certificates
undefined
Toxic
Health
Ergocalciferol (vitamin D2, calciferol) 97+%

Specification Test Results

  • Appearance:
    White to light yellow Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    113 to 119 °C
  • HPLC:
    97.0 to 103.0 %
  • Specific optical rotation:
    +100° to +106° (20 °C, 589 nm) (c=1,5; EtOH)

Specification Test Results

Related Information

Frequently Bought Together