Bromoxylenol blue, Sigma-Aldrich®
Catalog # 205478-10G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
EINECS:254-780-4
CAS Number:40070-59-5
Formula:C₂₃H₂₀Br₂O₅S
MDL Number:MFCD00005873