Bromoxylenol blue
Catalog # MOLEM34612776-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
EINECS:254-780-4
CAS Number:40070-59-5
Formula:C₂₃H₂₀Br₂O₅S
MDL Number:MFCD00005873