Brass (alloy 260), gauze woven from wire 100 mesh woven from 0.11mm (0.0045in)
Catalog # 45208.HB
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1886
- Pk Info:300×300 mm
Specifications
About this item
:
Storage Temperature:Ambient
MDL Number:MFCD00171255