Order Entry
Northern Ireland
ContactUsLinkComponent
Brass (alloy 260) (Cu:Zn; 70:30%) (w/w), gauze woven from wire 80 mesh, Ø 0.14 mm (0.0055 in)
Brass (alloy 260) (Cu:Zn; 70:30%) (w/w), gauze woven from wire 80 mesh, Ø 0.14 mm (0.0055 in)
Catalog # 45187.HB
CAS Number:  
Brass (alloy 260) (Cu:Zn; 70:30%) (w/w), gauze woven from wire 80 mesh, Ø 0.14 mm (0.0055 in)
Catalog # 45187.HB
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1886
  • Pk Info:
    300×300 mm

Specifications

About this item

 :
Storage Temperature:Ambient
MDL Number:MFCD00171255