D-(+)-Glucurono-6,3-lactone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:32449-92-6
:
Formula:C₆H₈O₆
MDL Number:MFCD00135622
:176.12 g/mol
:M-Clarity™ quality level = MQ200
EINECS:251-053-3
Specification Test Results
- Melting Point:170 - 175 °C
- Molar Mass:176.12 g/mol
Specification Test Results
Frequently Bought Together