Order Entry
Northern Ireland
ContactUsLinkComponent
4-(2-Pyrrolidinyl)pyridine 96%
Irritant
4-(2-Pyrrolidinyl)pyridine 96%
Catalog # H26051.MD
CAS Number:  
Irritant
4-(2-Pyrrolidinyl)pyridine 96%
Catalog # H26051.MD
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

CAS Number:128562-25-4
 :
Formula:C₉H₁₂N₂
MDL Number:MFCD01862536
 :148.21 g/mol
 :GHS07
Storage Temperature:Ambient