

4-(1-Pyrrolidinyl)pyridine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2456-81-7
:
Formula:C₉H₁₂N₂
MDL Number:MFCD00006415
:148.21 g/mol
Boiling Pt:171…173 °C (21 mmHg)
Melting Pt:54…58 °C
Flash Pt:142 °C (287 °F)
Storage Temperature:Ambient
EINECS:219-534-2
UN:2923
ADR:8,II
:GHS06
:GHS05
Frequently Bought Together