
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde ≥97%
Catalog # H57939.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:947-95-5
:
Formula:C₉H₁₁ClN₂O
MDL Number:MFCD00100755
:220.65 g/mol
Melting Pt:145…148 °C
:GHS07
Storage Temperature:Ambient