Order Entry
Northern Ireland
ContactUsLinkComponent
4,7,10-Trioxa-1,13-tridecanediamine for synthesis, Sigma-Aldrich®
4,7,10-Trioxa-1,13-tridecanediamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
4,7,10-Trioxa-1,13-tridecanediamine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Melting Point:
    -32 °C
  • Molar Mass:
    220.31 g/mol
  • Boiling Point:
    146 - 148 °C (1 hPa)
  • Vapor Pressure:
     - 0.001 hPa (20 °C)
  • Flash Point:
    139 °C
  • Explosion Limit:
    1.1 - 4.5 %(V)
  • Density:
    1.01 g/cm3 (20 °C)
  • pH:
    12 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    260 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together