
2-Ethylhexyl-4-(dimethylamino)benzoate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21245-02-3
:
Formula:C₁₇H₂₇NO₂
MDL Number:MFCD00017526
:277.40 g/mol
Merck Index:14,3232
Boiling Pt:325 °C
Density:0,995
Flash Pt:113 °C (235 °F)
EINECS:244-289-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together