HPLC columns, Hypersil GOLD™ C4
Supplier: Thermo Fisher Scientific
Certificates
About this item
These columns feature lower hydrophobicity than C18 or C8 and are recommended for very hydrophobic analytes.
- Ideal for analysis of hydrophobic polypeptides and proteins
- Highly symmetrical peak shapes
- 1,9 µm particle size columns improve speed and efficiency
Frequently Bought Together