Order Entry
Northern Ireland
ContactUsLinkComponent
Petri dish electrode
Petri dish electrode
  732-0000
 :  BTX
undefined
Petri dish electrode
  732-0000
 :  BTX

 

  • Description:
    Petri dish electrode, model 366, stainless steel, 2 mm gap, for 100 mm petri dish
  • Pk:
    1

 

 

This petri dish electrode is designed to be used with a 100 mm petri dish that functions as the electroporation chamber. The electrode is used to electroporate adherent cells or tissue grown in a petri dish.

  • Easy to use: Users simply need to add the exogenous molecule of interest into the electroporation buffer
  • Electrode assembly contains parallel stainless steel electrodes which generate a homogeneous field
  • Petri dish electrode is compatible with most BTX Generators

Typical applications include adherent mammalian cell transfections and plant tissue cell transfections.

Generator compatibility: ECM 830, ECM 630 and ECM 2001
Voltage range: 0 to 2000 V

Pulse length range: 10 µsec to 10 msec

Volume range: 10 to 50 ml

Electrode gap: 2 mm