Adenosine-5'-triphosphate disodium salt hydrate 98%, Ultrapure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51963-61-2
:
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
:551.2 g/mol (anhy)
Merck Index:14,155
Storage Temperature:Freezer
EINECS:213-579-1
Related Information
Frequently Bought Together