Azure C
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:531-57-7
:
Formula:C₁₃H₁₂ClN₃S
MDL Number:MFCD00067677
:277.78 g/mol
Merck Index:14,929
Storage Temperature:Ambient
EINECS:208-512-8
Related Information
Frequently Bought Together