
Streptozocin ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18883-66-4
:
Formula:C₈H₁₅N₃O₇
MDL Number:MFCD00006607
:265.2 g/mol
Merck Index:14,8832
Melting Pt:121 °C
EINECS:242-646-8
:GHS08
Storage Temperature:Freezer
Related Information
Frequently Bought Together