α-Ketoglutaric acid disodium salt dihydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:305-72-6
:1282616-74-3
Formula:C₅H₄O₅·2Na·2H₂O
MDL Number:MFCD00150702
:226.092 g/mol
Storage Temperature:Ambient
EINECS:206-167-8
Related Information
Frequently Bought Together