Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Potassium chloride

Potassium chloride

Glass bottle for solids, 1 kg
Catalog # RC-124
Supplier:   G-Biosciences
Potassium chloride
Catalog # RC-124
Supplier:  G-Biosciences
CAS Number:  7447-40-7
Potassium chloride

Glass bottle for solids, 1 kg

CAS Number:  7447-40-7
Price
£107.00
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Potassiumchloride
  • CAS Number:7447-40-7
  • Formula:KCl
  • MDL Number:MFCD00011360
  • Molecular Weight:74.55 g/mol
  • Merck Index:14,07621
  • Boiling Pt:1420 °C (1013 hPa)
  • Melting Pt:773 °C
  • Density:1,98 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-211-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.