Bismarck brown R
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5421-66-9
:1052-38-6
Formula:C₂₁H₂₄N₈·2HCl
MDL Number:MFCD00036386
:461.39 g/mol
Melting Pt:222 °C, (dec.)
Storage Temperature:Ambient
EINECS:226-541-4
Related Information
Frequently Bought Together