Colchicine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64-86-8
:
Formula:C₂₂H₂₅NO₆
MDL Number:MFCD00078484
:399.44 g/mol
Merck Index:14,2471
Melting Pt:∼ 150 °C (dec.)
Density:1,32 g/cm³ (20 °C)
UN:1544
ADR:6.1,I
Related Information
Frequently Bought Together