L-Alanine cell culture reagent
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-41-7
:
Formula:C₃H₇NO₂
MDL Number:MFCD00064410
:89.09 g/mol
Merck Index:14,204
Boiling Pt:189 °C (1013 hPa)
Melting Pt:∼ 295 °C (dec.)
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-273-8
Related Information
Frequently Bought Together