Polyvinylpyrrolidone, M.W. ∼ 10,000 (K15)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9003-39-8
:
Formula:(C₆H₉NO)n
MDL Number:MFCD01076626
Merck Index:14,7697
Storage Temperature:Ambient
Related Information
Frequently Bought Together