
2-Aminopurine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:452-06-2
:
Formula:C₅H₅N₅
MDL Number:MFCD00005566
:135.13 g/mol
Melting Pt:280…282 °C
EINECS:207-197-4
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together