Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Thiomersal

Thiomersal

Toxic
Health
Environment
Thiomersal
Certificates
Thiomersal

2 Options Available Below

Product Details & Documents
Synonyms:  Ethylmercurithiosalicylic acid sodium salt, Sodium ethylmercurithiosalicylate, Thimerosal, Merthiolat, 2-[(Ethylmercury)thio]benzoic acid sodium salt, 2-(Ethylmercuriomercapto)benzoic acid sodium salt, 2-(Ethylmercuriothio)benzoic acid sodium salt
  • CAS Number:54-64-8
  • Formula:2-(C₂H₅HgS)C₆H₄CO₂Na
  • MDL Number:MFCD00013062
  • Molecular Weight:404.82 g/mol
  • Merck Index:14,9317
  • Melting Pt:234…237 °C
  • Flash Pt:250 °C (482 °F)
  • Storage Temperature:Ambient
  • EINECS:200-210-4
  • UN:2025
  • ADR:6.1,II
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - £130.00
    £130.00
    1 * 10 g
       £130.00

    Specifications:
    More
    Specifications
    Cat. No.
    J61799.09
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £228.00
    £228.00
    1 * 25 g
       £228.00

    Specifications:
    More
    Specifications
    Cat. No.
    J61799.14
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.