

Lithium hexafluoroantimonate ≥97%
Catalog # 38595.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:242.68 g/mol
Boiling Pt: Dec.
Storage Temperature:Ambient
UN:1549
ADR:6.1,III
:GHS07
:GHS09
CAS Number:18424-17-4
Formula:F₆LiSb
MDL Number:MFCD00042319