Order Entry
Northern Ireland
ContactUsLinkComponent
3'-Fluoro-4'-(trifluoromethyl)acetophenone ≥97%
Irritant
3'-Fluoro-4'-(trifluoromethyl)acetophenone ≥97%
Catalog # B22874.06
CAS Number:  
Irritant
3'-Fluoro-4'-(trifluoromethyl)acetophenone ≥97%
Catalog # B22874.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :206.14 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:237761-81-8
Formula:C₉H₆F₄O
MDL Number:MFCD00236273