Order Entry
Northern Ireland
ContactUsLinkComponent
4-(Hexyloxy)phenylboronic acid 96%
4-(Hexyloxy)phenylboronic acid 96%
Catalog # H52874.06
CAS Number:  
4-(Hexyloxy)phenylboronic acid 96%
Catalog # H52874.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:121219-08-7
 :
Formula:C₁₂H₁₉BO₃
MDL Number:MFCD04039027
 :222.09 g/mol
 :GHS07
Storage Temperature:Ambient