Order Entry
Northern Ireland
ContactUsLinkComponent
 
3-Bromo-4-fluorobenzylamine hydrochloride 98%
Catalog # A19280.06
CAS Number:  
3-Bromo-4-fluorobenzylamine hydrochloride 98%
Catalog # A19280.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

CAS Number:202865-68-7
 :
Formula:C₇H₈BrClFN
MDL Number:MFCD00143094
 :240.50 g/mol
Melting Pt:214…218 °C
 :GHS07
Storage Temperature:Ambient