Divinyl sulphone stabilised for synthesis, Sigma-Aldrich®
Catalog # 8.21215.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 ml
- Seq:2275
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-26 °C
- Molar Mass:118.16 g/mol
- Boiling Point:234.3 °C (1013 hPa)
- Vapor Pressure:1.05 hPa (25 °C)
- Flash Point:102 °C
- Refractive Index:1.473 (27 °C, 589 nm)
- Density:1.18 g/cm3 (20 °C)
Specification Test Results
About this item
:
:118.16 g/mol
ADR:6.1 (8),I
:M-Clarity™ quality level = MQ200
:Stabilised with hydroquinone (HQ)
EINECS:201-057-6
UN:2927
:GHS06
:GHS05
Storage Temperature:Refrigerator
CAS Number:77-77-0
Formula:C₄H₆O₂S
MDL Number:MFCD00008623